missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, ≥99%, ReagentPlus™, Honeywell Fluka™

Product Code. 15682070
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
Packaging:
Drum with inliner
Poly Bottle

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Quantity 12 kg
Linear Formula NaBr
UN Number NONH for all modes of transport
Solubility Information Soluble in ethanol, Soluble in water
IUPAC Name sodium;bromide
Formula Weight 102.89g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Packaging Drum with inliner
Vapor Pressure 1mmHg (806°C)
Chemical Name or Material Sodium Bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'