missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 13225423
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Physical Form Crystals
Quantity 500 g
IUPAC Name sodium;bromide
Formula Weight 102.89
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 3.21kg
Chemical Name or Material Sodium Bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'