missing translation for 'onlineSavingsMsg'
Learn More
Sodium butyrate, 98+%, Thermo Scientific Chemicals Product Code.: 11423980

Sodium butyrate, 98+%, Thermo Scientific Chemicals

Product Code. 11423980
5 g
Click to view available options
Quantity:
5 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11423980

Brand: Thermo Scientific Alfa Aesar A11079.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone ac
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-54-7
Molecular Formula C4H7NaO2
Molecular Weight (g/mol) 110.09
MDL Number MFCD00002816
InChI Key MFBOGIVSZKQAPD-UHFFFAOYSA-M
Synonym sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1Show MoreShow Less
PubChem CID 5222465
ChEBI CHEBI:64103
SMILES [Na+].CCCC([O-])=O

Specifications

Melting Point >300°C
Linear Formula CH3(CH2)2CO2Na
Quantity 5 g
Beilstein 3629439
Sensitivity Hygroscopic
Solubility Information Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in ether.
IUPAC Name sodium butanoate
Formula Weight 110.09
Percent Purity ≥98%
Chemical Name or Material Sodium butyrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251854 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog NumberA11079.36,A11079.06,A11079.22
Lot NumberZ15J007 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog NumberA11079.36,A11079.06,A11079.22
Lot NumberY09I018 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog NumberA11079.36,A11079.06,A11079.22
Lot NumberR09I022 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA11079.36,A11079.06,A11079.22
Lot Number10239293 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11079.36,A11079.06,A11079.22
compliance-icons
Product Identifier
  • Sodium butyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Butanoic acid, sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium butyrate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.