Learn More
Sodium butyrate, 98+%
Beskrivelse
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.
Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Tekniske data
Tekniske data
| Melting Point | >300°C |
| Assay Percent Range | >98% |
| Quantity | 100 g |
| Beilstein | 3629439 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in ether. |
| IUPAC Name | sodium;butanoate |
| Formula Weight | 110.09 |
| Chemical Name or Material | Sodium butyrate |
Frequently Asked Questions (FAQs)
Sodium butyrate is soluble in water (0.1 g/mL), DMSO (<1 mg/mL), ethanol (~5 mg/mL), and PBS pH7. 2 (~10 mg/mL). Sodium butyrate is insoluble in ether.
Sikkerhed og håndtering
- Sodium butyrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Butanoic acid, sodium salt
RUO – Research Use Only
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.