Learn More
Sodium carbonate, 98%, Thermo Scientific Chemicals
£20.06 - £83.42
Chemical Identifiers
CAS | 497-19-8 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD00003494 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay sodaShow More |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317886
|
Thermo Scientific Alfa Aesar
L13098.36 |
500 g |
£20.06
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327886
|
Thermo Scientific Alfa Aesar
L13098.0E |
2500 g |
£58.01
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307886
|
Thermo Scientific Alfa Aesar
L13098.0I |
5000 g |
£83.42
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.
Solubility
Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide, acetone, and alkyl acetates.
Notes
m.p. 851° (begins to lose CO{2} at 400°)It is hygroscopic in nature, and absorbs moisture from air. On heating, it begins to lose carbon dioxide at about 400 °C.
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
[Na+].[Na+].[O-]C([O-])=O |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay sodaShow More | |
CHEBI:29377 |
Specifications
497-19-8 | |
CNa2O3 | |
4154566 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 | |
105.99 | |
Odorless | |
Hygroscopic | |
Sodium carbonate |
11.5 | |
MFCD00003494 | |
14,8596 | |
Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide,acetone,and alkyl acetates. | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
98% | |
1.535 | |
2.53 |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P234-P264b-P280i-P305+P351+P338-P390
H319
missing translation for 'einecsNumber' : 207-838-8
missing translation for 'rtecsNumber' : VZ4050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium carbonate, 98%, Thermo Scientific
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Disodium carbonate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.