Learn More
Sodium carbonate, 99.6%, ACS reagent, anhydrous, Thermo Scientific Chemicals
£29.25 - £93.77
Chemical Identifiers
CAS | 497-19-8 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD00003494 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
IUPAC Name | disodium carbonate |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10577182
|
Thermo Scientific Acros
424285000 |
500 g | Plastic bottle |
£29.25
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10237813
|
Thermo Scientific Acros
424280025 |
2.5 kg | Plastic bottle |
£93.77
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentChemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
disodium carbonate |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 | |
[Na+].[Na+].[O-]C([O-])=O |
Specifications
497-19-8 | |
Fine Powder | |
CNa2O3 | |
MFCD00003494 | |
15, 8731 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
105.99 | |
ACS Reagent | |
Sodium carbonate, Anhydrous |
White | |
99.6% | |
Na2CO3 | |
07,332 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
≥99.5% (on dried substance) | |
2.53 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 207-838-8
missing translation for 'rtecsNumber' : VZ4050000
missing translation for 'tsca' : TSCA
- Sodium carbonate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.