missing translation for 'onlineSavingsMsg'
Learn More
Sodium carbonate, 99.8%, for analysis, anhydrous, powder, Thermo Scientific Chemicals Product Code.: 10266690

Sodium carbonate, 99.8%, for analysis, anhydrous, powder, Thermo Scientific Chemicals

Product Code. 10266690
1 kg, Plastic Bottle
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10266690

Brand: Thermo Scientific Acros 207810010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Boiling Point 1600.0°C
Melting Point 851.0°C
Color White
Physical Form Crystalline Powder or Granules
Quantity 1 kg
Assay Percent Range 99.8%
Linear Formula Na2CO3
Fieser 7332
Merck Index 15, 8731
Solubility Information Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 105.99
Percent Purity 99.8%
Grade Analytical
Loss on Drying 0.5% max. (300°C)
Packaging Plastic Bottle
Chemical Name or Material Sodium carbonate, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464173 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number207810010,207810050
Lot NumberA0449649 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog Number207810010,207810050
Lot NumberA0394315 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number207810010,207810050
Lot NumberA0408780 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number207810010,207810050
Lot NumberA0405636 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number207810010,207810050
compliance-icons
Product Identifier
  • Sodium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium carbonate, 99.8%, for analysis, anhydrous, powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.