missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, 99.95%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog Number 10206410
44.44 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
10340
CHEBI:29377
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
10340
CHEBI:29377
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY
Boiling Point 1600.0°C
Melting Point 851.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 99.95%
Linear Formula Na2CO3
Fieser 07,332
Merck Index 15,8731
Solubility Information Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 105.99
Percent Purity 99.95%
Grade Extra Pure
Loss on Drying 1% (285°C) max.
Packaging Plastic bottle
Chemical Name or Material Sodium carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'