Learn More
Sodium carbonate, anhydrous, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
£51.20 - £1735.00
Chemical Identifiers
CAS | 497-19-8 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.99 |
MDL Number | MFCD00003494 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
SMILES | [Na+].[Na+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347017
|
Thermo Scientific Alfa Aesar
010861.06 |
5 g |
£51.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367017
|
Thermo Scientific Alfa Aesar
010861.14 |
25 g |
£163.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357017
|
Thermo Scientific Alfa Aesar
010861.22 |
100 g |
£493.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337017
|
Thermo Scientific Alfa Aesar
010861.36 |
500 g |
£1735.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
[Na+].[Na+].[O-]C([O-])=O |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 |
Specifications
497-19-8 | |
Powder | |
CNa2O3 | |
4154566 | |
Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide,acetone,and alkyl acetates. | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
Odorless | |
Hygroscopic | |
2.53 g/cm3 at 20°C |
White | |
99.997% (metals basis) | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
105.99 | |
1.535 | |
White powder | |
Sodium carbonate, anhydrous |
Safety and Handling
P234-P264b-P280i-P305+P351+P338-P390
H319
missing translation for 'einecsNumber' : 207-838-8
missing translation for 'rtecsNumber' : VZ4050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only