missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10162540 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2.5kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10162540

Brand: Fisher Chemical S/2880/65

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium Carbonate Anhydrous, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Boiling Point 1600°C
Melting Point 854°C
Color White
pH 11.3
Physical Form Solid
Quantity 5 kg
IUPAC Name disodium carbonate
Formula Weight 105.99g/mol
Packaging Plastic keg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.