missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Anhydrous, ULTREX™ Ultrapure Reagent, J.T. Baker™

Product Code. 15508324
Click to view available options
Quantity:
25 g
100 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Quantity 100 g
Formula Weight 105.99
Grade ULTREX™ Ultrapure Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.53kg
Chemical Name or Material Sodium Carbonate, Anhydrous
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'