missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Decahydrate, Certified AR for Analysis, Fisher Chemical™

£62.20 - £384.00

Chemical Identifiers

CAS 6132-02-1
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.14
MDL Number 149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10396190
View Documents
Fisher Chemical
S/2760/53
500 g Plastic powder jar This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10655502
View Documents
Fisher Chemical
S/2760/60
1 kg Plastic powder jar This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10000340
View Documents
Fisher Chemical
S/2760/65
5 kg Plastic keg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Sodium Carbonate Decahydrate, Certified AR for Analysis

Specifications

Chemical Identifiers

6132-02-1
286.14
XYQRXRFVKUPBQN-UHFFFAOYSA-L
151402
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
CH20Na2O13
149178
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
disodium;carbonate;decahydrate

Specifications

34°C
White
Solid
149178
XYQRXRFVKUPBQN-UHFFFAOYSA-L
disodium;carbonate;decahydrate
151402
6132-02-1
11 to 12
CH20Na2O13
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
286.14
286.01g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium carbonate decahydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Sodium carbonate decahydrate, Sodium carbonate

SDS
Documents

Documents

Product Certifications