missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate decahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.), Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
1 kg
2.5 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Max % | 100.0000% |
Quantity | 50 kg |
Assay Percent Range | 99.5 to 100.5% |
Linear Formula | Na2CO3 · 10H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 286.14 |
Percent Purity | 99.5 to 100.5% (ex dried subst.) |
Grade | puriss. |
Packaging | Fiberboard Box |
Density | 1.460 g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'