missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Monohydrate, Crystal, N.F. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
Physical Form | Crystals |
Quantity | 2.5 kg |
Formula Weight | 124 |
Grade | FCC/NF |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.25kg |
Chemical Name or Material | Sodium Carbonate, Monohydrate |
Safety and Handling
Product Identifier
- Sodium carbonate monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Sodium carbonate monohydrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'