missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Physical Form | Anhydrous |
Quantity | 1 kg |
Linear Formula | Na2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4154566 |
IUPAC Name | disodium;carbonate |
Formula Weight | 105.99g/mol |
Percent Purity | ≥99.8% (calc. to the dried substance) |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Sodium Carbonate, Reag. Ph. Eur. reag. ISO |
Safety and Handling

Product Identifier
- Sodium carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium carbonate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance), Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction