missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium Caseinate Pract.

Sodium caseinate, CAS # 9005-46-3, is the salt of the phosphoprotein class found naturally in milk and cheese. It contains all nine essential amino acids and may be used as a molecular reagent, or in cell culture research on growth and differentiation.

Chemical Identifiers

CAS 9005-46-3
MDL Number MFCD00130736
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Sodium caseinate is the salt of the phosphoprotein class known as caseins
• Caseins are found in animal tissues, especially milk, and they contain all nine essential amino acids
Applications
• Casein can be used in protease enzyme activity studies, in vitro experiments on cell growth and differentiation, or as a blocking solution for western blot
Specifications

Chemical Identifiers

9005-46-3
MFCD00130736

Specifications

9005-46-3
6 to 7.5 (5% soln.)
MFCD00130736
Powder
White to Cream
Authentic
≥92% (protein) (on dry substance)
Sodium caseinate, pract.
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : FI3540000

missing translation for 'tsca' : TSCA

Product Identifier
  • Sodium caseinate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only