Learn More
Sodium chlorate, 99+%, extra pure, Thermo Scientific™
Chemical Identifiers
CAS | 7775-09-9 |
---|---|
Molecular Formula | ClNaO3 |
Molecular Weight (g/mol) | 106.44 |
InChI Key | YZHUMGUJCQRKBT-UHFFFAOYSA-M |
Synonym | sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal |
PubChem CID | 516902 |
ChEBI | CHEBI:65242 |
IUPAC Name | sodium;chlorate |
SMILES | [O-]Cl(=O)=O.[Na+] |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Oxidation reagentChemical Identifiers
7775-09-9 | |
106.44 | |
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikalShow More | |
CHEBI:65242 | |
[O-]Cl(=O)=O.[Na+] |
ClNaO3 | |
YZHUMGUJCQRKBT-UHFFFAOYSA-M | |
516902 | |
sodium;chlorate |
Specifications
7775-09-9 | |
ClNaO3 | |
01,1056 | |
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal | |
YZHUMGUJCQRKBT-UHFFFAOYSA-M | |
sodium;chlorate | |
516902 | |
106.44 | |
Extra Pure | |
2.4900g/mL |
99+% | |
NaClO3 | |
15,8733 | |
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and glycerin,7.7g/L ethanol (20°C) | |
[O-]Cl(=O)=O.[Na+] | |
106.44 | |
CHEBI:65242 | |
99+% | |
2.49 | |
Sodium chlorate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
May cause fire or explosion; strong oxidizer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-887-4
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.