missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium chloride, 99.85%, for molecular biology, DNAse, RNAse and Protease free
Click to view available options
Quantity:
1 kg
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Boiling Point | 1461.0°C |
Melting Point | 801.0°C |
Color | White |
Physical Form | Crystalline Solid |
Quantity | 1 kg |
Assay Percent Range | 99.8% min. (Argentometry) |
Linear Formula | NaCl |
Fieser | 13,280 |
Merck Index | 15, 8734 |
Solubility Information | Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'