Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals
Sodium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals
Product Code. 10577372Product Code. 10577372
Brand: Thermo Scientific Acros 424290010

Use promo code "25339" to get your promotional price.
Explore more special offers
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Chemical Identifiers
CAS | 7647-14-5 |
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Specifications
Boiling Point | 1461.0°C |
Melting Point | 801.0°C |
Color | Brown-Yellow to Orange |
pH | 5.0 to 9.0 (5% soln. at 25°C) |
Physical Form | Adhering Powder and/or Lumps |
Quantity | 1 kg |
Assay Percent Range | 99+% |
Linear Formula | NaCl |
Fieser | 13,280 |
Merck Index | 15, 8734 |
Solubility Information | Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid |
IUPAC Name | sodium chloride |
Formula Weight | 58.44 |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Identification | Pass Test |
Loss on Drying | ≤0.5% |
Packaging | Amber Glass Bottle |
Vapor Pressure | 1mmHg at 440°C |
Chemical Name or Material | Sodium chloride |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
-
Product Code. 10428420
-
-
Product Code. 15638170
-
Product Code. 15648140
-
Product Code. 10092740
-
-
Product Code. 15646770
-
Product Code. 10055850
-
Product Code. 10478283
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
-
Catalog No. 10092740
£21.51 / 1kg -
Catalog No. 10112640
£50.40 / 1kg -
Catalog No. 10316943
£56.15 / 1kg -
Catalog No. 10428420
£72.05 / 1kg -
Catalog No. 12675077
£38.08 / 1kg -
Catalog No. 15526005
£32.15 / 500g -
Catalog No. 11482874
£111.00 / 2.50 L -
Catalog No. 10703811
£258.00 / 500g -
Catalog No. 10793611
£101.00 / 1kg -
Catalog No. 10117113
£25.71 / 100g -
Catalog No. 10401841
£22.81 / 500mL -
Catalog No. 15681810
£34.55 / 25g
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Lot Number2497961 | Certificate TypeCertificate of Analysis | Date31/01/2025 | Catalog Number424290050,424290251,424295000,424290250,424290010 |
Lot Number2407894 | Certificate TypeCertificate of Analysis | Date31/01/2025 | Catalog Number424290050,424290251,424295000,424290250,424290010 |
Lot Number2404508 | Certificate TypeCertificate of Analysis | Date31/01/2025 | Catalog Number424290050,424290251,424295000,424290250,424290010 |
Lot Number2349711 | Certificate TypeCertificate of Analysis | Date27/01/2025 | Catalog Number424290050,424290251,424295000,424290250,424290010 |
Lot Number2192435 | Certificate TypeCertificate of Analysis | Date27/01/2025 | Catalog Number424290050,424290251,424295000,424290250,424290010 |
- 1
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Hot Deal*Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.