Learn More
Sodium chloride, ACS, 99.0% min, Thermo Scientific Chemicals
Description
Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1413°C |
Melting Point | 801°C |
Physical Form | Crystalline |
Quantity | 500 g |
Assay Percent Range | 99.0% min |
Solubility Information | Soluble in water,glycerol. Very slightly soluble in alcoholFreely soluble in water. Slightly soluble in alcohol and ammonia. |
IUPAC Name | sodium chloride |
Formula Weight | 58.44 |
Grade | ACS Reagent |
Refractive Index | 1.5442 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N27L031 | Certificate of Analysis | 03/03/2025 | 012314.A9,012314.36,012314.A3 |
N27L001 | Certificate of Analysis | 28/02/2025 | 012314.A9,012314.36,012314.A3 |
N18L061 | Certificate of Analysis | 20/02/2025 | 012314.A9,012314.36,012314.A3 |
2404508 | Certificate of Analysis | 31/01/2025 | 012314.A9,012314.36,012314.A3 |
2497961 | Certificate of Analysis | 31/01/2025 | 012314.A9,012314.36,012314.A3 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.