missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, crystal, BAKER ANALYZED™, J.T. Baker™, Avantor™

Product Code. 10284640
Click to view available options
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Boiling Point 1413°C (1013 hPa)
Melting Point 801°C
CAS 7647-14-5
pH 5.0 to 8.0 (5% Solution at 25°C)
Quantity 1 kg
Assay Percent Range min. 99.5%
Molecular Formula ClNa
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Packaging Plastic bottle for solids
Density 2.16 g/cm³ (20°C)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'