missing translation for 'onlineSavingsMsg'
Learn More
Sodium Chloride, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10356340

Sodium Chloride, Extra Pure, SLR, Fisher Chemical™

Product Code. 10356340
5 kg
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
This item is not returnable. View return policy

Product Code. 10356340

Brand: Fisher Chemical S/3120/65

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 5 kg
IUPAC Name sodium;chloride
Formula Weight 58.44g/mol
Packaging Plastic powder jar
Vapor Pressure 1mmHg 865

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527965 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberS/3120/60,S/3120/70,S/3120/53,S/3120/63,S/3120/60X,S/3120/65
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Chloride, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.