missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, Extra Pure, SLR, Fisher Chemical
28.58 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Specifications
Boiling Point | 1461°C |
Melting Point | 801°C |
CAS | 7647-14-5 |
Color | White |
pH | 5.0 to 8.0 |
Physical Form | Solid |
Quantity | 1 kg |
Molecular Formula | ClNa |
MDL Number | MFCD00003477 |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'