Learn More
Sodium chloride, extra pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification
Spécification
Boiling Point | 1461.0°C |
Melting Point | 801.0°C |
Color | White |
pH | 5.0 to 8.0 (5%, H<2O, at 20°C) |
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Linear Formula | NaCl |
Fieser | 13,280 |
Merck Index | 15, 8734 |
Solubility Information | Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid |
Afficher plus |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463601 | Certificate of Analysis | 06/06/2024 | 194090250,194090050,194090010 |
A0456519 | Certificate of Analysis | 21/09/2023 | 194090250,194090050,194090010 |
A0456518 | Certificate of Analysis | 21/09/2023 | 194090250,194090050,194090010 |
A0456489 | Certificate of Analysis | 21/09/2023 | 194090250,194090050,194090010 |
A0456484 | Certificate of Analysis | 21/09/2023 | 194090250,194090050,194090010 |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.