missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, extra pure, Thermo Scientific Chemicals
£18.69 - £216.08
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10127853
|
Thermo Scientific Acros
194090010 |
1 kg | Plastic Bottle |
£18.69
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10287903
|
Thermo Scientific Acros
194090050 |
5 kg | Plastic Bottle |
£68.84
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10085673
|
Thermo Scientific Acros
194090250 |
25 kg | Fibre drum |
£216.08
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 |
Specifications
1461.0°C | |
7647-14-5 | |
5.0 to 8.0 (5%, H<2O, at 20°C) | |
ClNa | |
MFCD00003477 | |
15, 8734 | |
Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
≥99.0% | |
Sodium chloride |
801.0°C | |
White | |
Crystalline Powder | |
NaCl | |
13,280 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44 | |
Extra Pure |
Safety and Handling
missing translation for 'einecsNumber' : 231-598-3
RUO – Research Use Only