missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, Honeywell Fluka™
Click to view available options
Quantity:
500 g
5 kg
10 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Crystalline |
Quantity | 5 kg |
Percent Purity | ≥98% |
Packaging | Plastic Bottle |
Appearance | Complying |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'