missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Product Code. 15633410 Shop All Honeywell Fluka Products25 kg
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
25 kg
4 x 5 kg
6 x 1 kg
This item is not returnable.
View return policy
Product Code. 15633410
Brand: Honeywell-Fluka 3143425KG
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Chemical Identifiers
CAS | 7647-14-5 |
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Specifications
Boiling Point | 1,465°C |
Melting Point | 801°C (literature) |
CAS Max % | ≤100.0000% |
Quantity | 25 kg |
Linear Formula | NaCl |
UN Number | NONH for all modes of transport |
Beilstein | 3534976 |
Solubility Information | Soluble in water, 100mg/mL |
IUPAC Name | sodium chloride |
Formula Weight | 58.44 |
Percent Purity | >99.5% |
Grade | ACS Reagent/Reagent ISO/Reagent Ph. Eur. |
Packaging | Paper / Poly Bag |
Density | 2.160 g/cm3 |
Chemical Name or Material | Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
Savings of £134.86Product Code. 15698170
-
Savings of £109.30Product Code. 15668170
-
Savings of £59.18Product Code. 10514283
-
Product Code. 15653410
-
Product Code. 15691300
-
Product Code. 15673410
-
Product Code. 15683410
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 7
-
Catalog No. 10428420
£72.05 / 1kg -
Catalog No. 10703811
£258.00 / 500g -
Catalog No. 10793611
£101.00 / 1kg -
Catalog No. 10117113
£25.71 / 100g -
Catalog No. 10112640
£50.40 / 1kg -
Catalog No. 10092740
£21.51 / 1kg -
Catalog No. 10401841
£22.81 / 500mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction