missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%
£20.62 - £528.18
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15663410
|
Honeywell-Fluka
31434-500G |
500 g |
£20.62
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15623410
|
Honeywell-Fluka
31434-1KG |
1 kg |
£30.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15693410
|
Honeywell-Fluka
31434-6X500G |
6 x 500 g |
£111.34
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15673410
|
Honeywell-Fluka
31434-5KG |
5 kg |
£123.71
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15683410
|
Honeywell-Fluka
31434-6X1KG |
6 x 1 kg |
£163.38
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15653410
|
Honeywell-Fluka
31434-4X5KG |
4 x 5 kg |
£448.31
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15633410
|
Honeywell-Fluka
31434-25KG |
25 kg |
£528.18
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 |
Specifications
7647-14-5 | |
NaCl | |
NONH for all modes of transport | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44g/mol | |
ACS/puriss. p.a. |
ClNa | |
MFCD00003477 | |
3534976 | |
Soluble in water, 100mg/mL | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
≥99.5% | |
Sodium Chloride, Reag. Ph. Eur. reag. ISO |
Safety and Handling
missing translation for 'einecsNumber' : 231-598-3
missing translation for 'rtecsNumber' : VZ4725000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium chloride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™