missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, ReagentPlus™, Honeywell™

Reagent Grade,≥99%

£17.31 - £190.50

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15658140
View Documents
Honeywell Research Chemicals
S9625-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15648140
View Documents Promotion Details
Honeywell Research Chemicals
S9625-1KG
1 kg
£28.97
1kg
Please to purchase this item. Need a web account? Register with us today!
15668140
View Documents Promotion Details
Honeywell Research Chemicals
S9625-5KG
5 kg
£101.44
5kg
Please to purchase this item. Need a web account? Register with us today!
15638140
View Documents
Honeywell Research Chemicals
S9625-10KG
10 kg
£190.50
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
[Na+].[Cl-]
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710

Specifications

7647-14-5
NaCl
NONH for all modes of transport
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
58.44
CHEBI:26710
≥99%
Sodium chloride
ClNa
MFCD00003477
3534976
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44g/mol
ReagentPlus™
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : VZ4725000

SDS
Documents

Documents

Product Certifications