missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STE Buffer, Sodium Chloride-Tris-EDTA, 1X Solution, pH 8.0, Fisher BioReagents
£83.60
Specifications
Chemical Name or Material | Sodium Chloride-Tris-EDTA Buffer |
---|---|
Name Note | 1X, pH 8.0 |
CAS | 7647-14-5, 60-00-4, 7647-01-0, 7732-18-5 |
Boiling Point | 100°C |
Color | Undesignated |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10706844
|
Fisher BioReagents
BP2478-1 |
1 L | Nalgene Poly Bottle | ||||||
Description
STE buffer is suitable for biomolecular lab procedures.Specifications
Sodium Chloride-Tris-EDTA Buffer | |
7647-14-5, 60-00-4, 7647-01-0, 7732-18-5 | |
Undesignated | |
Liquid | |
Filtered through a 5-micron filter. |
1X, pH 8.0 | |
100°C | |
8 | |
DNase free | |
Protease free |
Safety and Handling
Recommended Storage : RT