Learn More
Sodium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
£65.40 - £310.00
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351289
|
Thermo Scientific Alfa Aesar
013683.06 |
5 g |
£65.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361289
|
Thermo Scientific Alfa Aesar
013683.14 |
25 g |
£310.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media, a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 |
Specifications
1413°C | |
7647-14-5 | |
6 to 9 | |
ClNa | |
14,8599 | |
Soluble in water (358mg/ml at 20°C),alcohol (very slightly),glycerol (100mg/ml),and ammonia. Insoluble in hydrochloric acid. | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
99.998% | |
Ampule | |
Hygroscopic | |
(metals basis) |
801°C | |
White | |
Beads | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium;chloride | |
5234 | |
58.44 | |
Odorless | |
1.5442 | |
2.165 g/mL | |
Sodium chloride, Ultra dry |
Safety and Handling
missing translation for 'einecsNumber' : 231-598-3
missing translation for 'rtecsNumber' : VZ4725000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only