missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chlorite, 80%, pure, unstabilized
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
2.5kg
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 180.0°C to 200.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 80% |
| Molecular Formula | ClNaO2 |
| Linear Formula | NaClO2 |
| Fieser | 05,603; 09,423; 11,481; 13,280 |
| Merck Index | 15, 8735 |
| Molecular Weight (g/mol) | 90.438 |
| Show More |
Safety and Handling
Product Identifier
- Sodium chlorite, unstabilized
- Danger
- Oxidising solid Category 1
- Acute toxicity Category 3
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H271-May cause fire or explosion; strong oxidiser.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : Sodium chlorite, Sodium carbonate, Sodium chlorate, Sodium hydroxide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction