missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chromate, anhydrous, 98%, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Sodium chromate anhydrous is used as a corrosion inhibitor in the petroleum industry, wood preservative and dyeing auxiliary in the textile industry. It is also used to prepare pigments for paints and inks. It also finds use in pharmaceuticals to determine red blood cell volume. In synthetic chemistry, it is utilized as an oxidant to convert primary alcohols and secondary alcohols to carboxylic acids and ketones respectively.
Solubility
Soluble in water. Slightly soluble in methanol and ethanol.
Notes
Hygroscopic. Incompatible with combustible materials and reducing agents.
Sodium chromate anhydrous is used as a corrosion inhibitor in the petroleum industry, wood preservative and dyeing auxiliary in the textile industry. It is also used to prepare pigments for paints and inks. It also finds use in pharmaceuticals to determine red blood cell volume. In synthetic chemistry, it is utilized as an oxidant to convert primary alcohols and secondary alcohols to carboxylic acids and ketones respectively.
Solubility
Soluble in water. Slightly soluble in methanol and ethanol.
Notes
Hygroscopic. Incompatible with combustible materials and reducing agents.
Specifications
Specifications
Melting Point | 792°C |
Quantity | 500 g |
UN Number | UN3288 |
Merck Index | 14,8601 |
Synonym | sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal |
Solubility Information | Soluble in water. Slightly soluble in methanol and ethanol. |
IUPAC Name | sodium;chlorate |
Formula Weight | 161.97 |
Percent Purity | 98% |
Odor | Odorless |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'