missing translation for 'onlineSavingsMsg'
Learn More

Sodium chromate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

For Use As Corrosion Inhibitor, in Pigment Manufacture

£42.21 - £138.69

Chemical Identifiers

CAS 10034-82-9
Molecular Formula CrH8Na2O8
Molecular Weight (g/mol) 234.032
MDL Number MFCD00149167
InChI Key AVHUVTPOXZEIRG-UHFFFAOYSA-N
Synonym sodium chromate tetrahydrate, cro4.2na.4h2o, acmc-20ajol, disodium chromate tetrahydrate, ksc492q3d, disodium chromate 2-tetrahydrate, sodium chromate tetrahydrate, reagent grade, sodium chromate tetrahydrate, p.a
PubChem CID 22146515
IUPAC Name disodium;dioxido(dioxo)chromium;tetrahydrate
SMILES O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396808
View Documents
Thermo Scientific Alfa Aesar
013453.30
250 g
£42.21
250g
Please to purchase this item. Need a web account? Register with us today!
11386808
View Documents
Thermo Scientific Alfa Aesar
013453.A1
1 kg
£138.69
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Corrosion inhibitor, pigment manufactureSodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10034-82-9
234.032
AVHUVTPOXZEIRG-UHFFFAOYSA-N
22146515
O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]
CrH8Na2O8
MFCD00149167
sodium chromate tetrahydrate, cro4.2na.4h2o, acmc-20ajol, disodium chromate tetrahydrate, ksc492q3d, disodium chromate 2-tetrahydrate, sodium chromate tetrahydrate, reagent grade, sodium chromate tetrahydrate, p.a
disodium;dioxido(dioxo)chromium;tetrahydrate

Specifications

10034-82-9
Crystals
MFCD00149167
Soluble in water.
O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]
234.032
234.04 (161.97 Anhydrous)
Odorless
Yellow crystalline
Sodium chromate tetrahydrate
Yellow
CrH8Na2O8
sodium chromate tetrahydrate, cro4.2na.4h2o, acmc-20ajol, disodium chromate tetrahydrate, ksc492q3d, disodium chromate 2-tetrahydrate, sodium chromate tetrahydrate, reagent grade, sodium chromate tetrahydrate, p.a
AVHUVTPOXZEIRG-UHFFFAOYSA-N
disodium;dioxido(dioxo)chromium;tetrahydrate
22146515
Reagent
Hygroscopic
2.73 g/cm3
Videos
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301-H312-H314-H317-H330-H334-H335-H340-H350-H360-H372

missing translation for 'dotInformation' : Hazard Class: 6.1, Packing Group: II

missing translation for 'einecsNumber' : 231-889-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only