missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate, Dihydrate, Granular, U.S.P. - F.C.C., J.T. Baker™
£140.00 - £330.00
Chemical Identifiers
CAS | 6132-04-3 |
---|---|
Molecular Formula | C6H9Na3O9 |
Molecular Weight (g/mol) | 294.10 |
InChI Key | NLJMYIDDQXHKNR-UHFFFAOYSA-K |
Synonym | trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate |
PubChem CID | 71474 |
ChEBI | CHEBI:32142 |
IUPAC Name | trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |
SMILES | C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15568154
|
Avantor J.T.Baker
3649-01 |
500 g | Wide Mouth Amber Glass Bottle |
£140.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15578154
|
Avantor J.T.Baker
3649-05 |
2.5 kg | Poly Bottle |
£330.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
6132-04-3 | |
294.10 | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
CHEBI:32142 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] |
C6H9Na3O9 | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
71474 | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |
Specifications
6132-04-3 | |
C6H9Na3O9 | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate | |
71474 | |
294.1 | |
1L = 1.70kg |
Granules | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] | |
294.10 | |
CHEBI:32142 | |
FCC/USP | |
Sodium Citrate, Dihydrate |