missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate Dihydrate (Small, Colorless Granules), Fisher BioReagents™
32.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Description
Sodium Citrate Dihydrate (Small, Colorless Granules)
Sodium Citrate is used primarily in the preparation of SSC buffer used in Southern transfers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Decomposition Information | >230°C |
Melting Point | 300°C |
Color | White |
pH | 7.5 to 9.5 |
Assay Percent Range | 99.0 to 101.0 % |
Packaging | Poly Bottle |
Quantity | 1 kg |
Merck Index | 15, 8737 |
Solubility Information | 0.005% max. Insoluble Matter |
Formula Weight | 294.09 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'