Learn More
Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1497.0°C |
Melting Point | 562.0°C |
Color | White |
Physical Form | Crystals |
Quantity | 5 g |
Assay Percent Range | 98% min. (Argentometry) |
Linear Formula | NaCN |
Beilstein | 02, IV, 50 |
Fieser | 04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185 |
Merck Index | 15, 8741 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468457 | Certificate of Analysis | 31/01/2025 | 370310025,370310010,370310050 |
A0465584 | Certificate of Analysis | 03/09/2024 | 370310025,370310010,370310050 |
A0465000 | Certificate of Analysis | 03/09/2024 | 370310025,370310010,370310050 |
A0457320 | Certificate of Analysis | 23/07/2024 | 370310025,370310010,370310050 |
A0457320 | Certificate of Analysis | 23/07/2024 | 370310025,370310010,370310050 |
Safety and Handling




- Sodium cyanide
- Danger
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 1
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH032-Contact with acids liberates very toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.