missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyclohexanebutyrate, AAS, Certified Na Content, Thermo Scientific™

Product Code. 11304969
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61886-29-1
Molecular Formula C10H17NaO2
Molecular Weight (g/mol) 192.234
MDL Number MFCD00036395
InChI Key GBEFSJZAXXWKIO-UHFFFAOYSA-M
Synonym sodium cyclohexanebutyrate, sodium 4-cyclohexylbutanoate, cyclohexanebutanoic acid, sodium salt, cyclohexanebutanoic acid, sodium salt 1:1, sodium 4-cyclohexylbutyrate, 4-cyclohexylbutyric acid sodium salt, 4-cyclohexylbutanoic acid, sodium salt, sodium cyclohexanebutyrate, aas, certified na content
PubChem CID 4072353
IUPAC Name sodium;4-cyclohexylbutanoate
SMILES C1CCC(CC1)CCCC(=O)[O-].[Na+]

Specifications

Melting Point 264°C (decomposition)
Quantity 0.5 g
Formula Weight 192.23
Grade AAS
Physical Form Powder
Chemical Name or Material Sodium cyclohexanebutyrate, Certified Na Content
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.