missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyclopentadienide, 2-3M in THF, Thermo Scientific Chemicals

Product Code. 11326389
Click to view available options
Quantity:
0.1 mol
0.5 mol

Reagent for the preparation of metallocenes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4984-82-1
Molecular Formula C5H5Na
Molecular Weight (g/mol) 88.085
MDL Number MFCD00012161
InChI Key JQUZTGJSSQCTPV-UHFFFAOYSA-N
Synonym cyclopentadienylsodium, sodium cyclopenta-1,3-dien-1-ide, sodium cyclopentadienide, 2.0m solution in tetrahydrofuran
PubChem CID 21915903
IUPAC Name sodium;cyclopenta-1,3-diene
SMILES C1C=CC=[C-]1.[Na+]

Specifications

Color Pink to Red
Density 0.938 g/mL
Flash Point −17°C (1°F)
Odor Ether-like
Quantity 0.1 mol
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 88.09
Concentration or Composition (by Analyte or Components) 2 to 3M in THF
Physical Form Liquid
Chemical Name or Material Sodium cyclopentadienide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'