missing translation for 'onlineSavingsMsg'
Learn More
Sodium D-gluconate, 97%, Thermo Scientific Chemicals Product Code.: 11498693

Sodium D-gluconate, 97%, Thermo Scientific Chemicals

Product Code. 11498693
2500 g
Click to view available options
Quantity:
2500 g
10 kg
This item is not returnable. View return policy

Product Code. 11498693

Brand: Thermo Scientific Alfa Aesar A10464.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.

Solubility
Very soluble in water; sparingly soluble in alcohol; insoluble in ether.

Notes
In general it is stable, non corrosive, non-toxic, readily biodegradable and incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.137
MDL Number MFCD00064210
InChI Key UPMFZISCCZSDND-JJKGCWMISA-M
Synonym sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100tShow MoreShow Less
PubChem CID 23672301
ChEBI CHEBI:84997
IUPAC Name sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]

Specifications

Melting Point ∽206°C (decomposition)
Quantity 2500 g
Beilstein 3919651
Solubility Information Very soluble in water; sparingly soluble in alcohol; insoluble in ether.
Formula Weight 218.14
Chemical Name or Material Sodium D-gluconate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN03L042 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberA10464.0E,A10464.0C
Lot NumberS11K008 Certificate TypeCertificate of Analysis Date19/06/2024 Catalog NumberA10464.0E,A10464.0C
Lot NumberT21J001 Certificate TypeCertificate of Analysis Date26/07/2023 Catalog NumberA10464.0E,A10464.0C
Lot NumberH16Z019 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10464.0E,A10464.0C
Lot NumberB18Z001 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10464.0E,A10464.0C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium D-gluconate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.