missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Diethyldithiocarbamate Trihydrate, 99+%, Extra Pure, SLR, Fisher Chemical™
£83.45 - £231.00
Chemical Identifiers
CAS | 20624-25-3 |
---|---|
Molecular Formula | C5H16NNaO3S2 |
Molecular Weight (g/mol) | 225.297 |
MDL Number | 150617 |
InChI Key | WWGXHTXOZKVJDN-UHFFFAOYSA-M |
Synonym | sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech |
PubChem CID | 517546 |
IUPAC Name | sodium;N,N-diethylcarbamodithioate;trihydrate |
SMILES | CCN(CC)C(=S)[S-].O.O.O.[Na+] |
Description
Chemical Identifiers
20624-25-3 | |
225.297 | |
WWGXHTXOZKVJDN-UHFFFAOYSA-M | |
517546 | |
CCN(CC)C(=S)[S-].O.O.O.[Na+] |
C5H16NNaO3S2 | |
150617 | |
sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech | |
sodium;N,N-diethylcarbamodithioate;trihydrate |
Specifications
95°C | |
White | |
Solid | |
150617 | |
WWGXHTXOZKVJDN-UHFFFAOYSA-M | |
sodium;N,N-diethylcarbamodithioate;trihydrate | |
517546 | |
Plastic powder jar |
20624-25-3 | |
7 | |
C5H16NNaO3S2 | |
sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech | |
CCN(CC)C(=S)[S-].O.O.O.[Na+] | |
225.297 | |
225.31g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Diethyldithiocarbamic acid, sodium salt trihydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Diethylcarbamodithioic acid sodium salttrihydrate, Sodium diethyldithiocarbamate