missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals

£18.89 - £228.00

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
MDL Number MFCD00149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11461880
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11316.22
100 g
£18.89
100g
Please to purchase this item. Need a web account? Register with us today!
11471880
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11316.36
500 g
£27.74
500g
Please to purchase this item. Need a web account? Register with us today!
15443037
View Documents
Thermo Scientific Alfa Aesar
A11316.0E
2500 g
£82.21
2500g
Please to purchase this item. Need a web account? Register with us today!
15433037
View Documents
Thermo Scientific Alfa Aesar
A11316.0C
10 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

Solubility
Soluble in water.(1 M at 20°C)

Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
Specifications

Chemical Identifiers

13472-35-0
MFCD00149209
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
sodium;dihydrogen phosphate;dihydrate
H6NaO6P
VBJGJHBYWREJQD-UHFFFAOYSA-M
23673460

Specifications

13472-35-0
NaH2PO4·2H2O
14,8660
Soluble in water.(1 M at 20°C)
O.O.OP(=O)(O)[O-].[Na+]
156.006
156.01 (119.98 Anhydrous)
Hygroscopic
Sodium dihydrogen phosphate dihydrate
H6NaO6P
MFCD00149209
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
VBJGJHBYWREJQD-UHFFFAOYSA-M
sodium;dihydrogen phosphate;dihydrate
23673460
99%
1.91
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-449-2

missing translation for 'rtecsNumber' : WA1900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Sodium dihydrogen phosphate dihydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium dihydrogen phosphate dihydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.