Learn More
Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Description
Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 100°C-H2O |
Color | White |
Physical Form | Crystalline |
Quantity | 1 kg |
Assay Percent Range | 98.0-102.0% |
Solubility Information | Highly soluble in water. Insoluble in ethanol,ether and chloroform. |
IUPAC Name | sodium phosphoric acid hydrate |
Formula Weight | 137.99 (119.98 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M07L005 | Certificate of Analysis | 10/01/2025 | 011591.30,011591.A1,011591.A7 |
W24K029 | Certificate of Analysis | 03/10/2024 | 011591.30,011591.A1,011591.A7 |
S03K015 | Certificate of Analysis | 11/06/2024 | 011591.30,011591.A1,011591.A7 |
N07K034 | Certificate of Analysis | 13/02/2024 | 011591.30,011591.A1,011591.A7 |
X05J045 | Certificate of Analysis | 24/10/2023 | 011591.30,011591.A1,011591.A7 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
16Minutes
40Seconds
57
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.