Learn More
Sodium diphenylamine-4-sulfonate, 98%, Thermo Scientific Chemicals
£47.94 - £161.81
Chemical Identifiers
CAS | 6152-67-6 |
---|---|
Molecular Formula | C12H11NNaO3S |
Molecular Weight (g/mol) | 272.27 |
MDL Number | MFCD00036468 |
InChI Key | VMVSDVIYIMNZBS-UHFFFAOYSA-N |
Synonym | sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt |
PubChem CID | 517002 |
IUPAC Name | sodium;4-anilinobenzenesulfonate |
SMILES | [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468637
|
Thermo Scientific Alfa Aesar
A19768.06 |
5 g |
£47.94
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478637
|
Thermo Scientific Alfa Aesar
A19768.14 |
25 g |
£161.81
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6152-67-6 | |
272.27 | |
VMVSDVIYIMNZBS-UHFFFAOYSA-N | |
517002 | |
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 |
C12H11NNaO3S | |
MFCD00036468 | |
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt | |
sodium;4-anilinobenzenesulfonate |
Specifications
6152-67-6 | |
MFCD00036468 | |
VMVSDVIYIMNZBS-UHFFFAOYSA-N | |
sodium;4-anilinobenzenesulfonate | |
517002 | |
98% |
C12H11NNaO3S | |
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt | |
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1 | |
272.27 | |
271.27 | |
Sodium diphenylamine-4-sulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 228-165-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium diphenylamine-4-sulfonate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Sodium diphenylamine-4-sulfonate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.