missing translation for 'onlineSavingsMsg'
Learn More

Sodium, dispersion, 40% in oil, Thermo Scientific Chemicals

£106.53 - £285.42

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331428
View Documents
Thermo Scientific Alfa Aesar
010340.30
250 g
£106.53
250g
Please to purchase this item. Need a web account? Register with us today!
11321428
View Documents
Thermo Scientific Alfa Aesar
010340.56
4 x 250 g
£285.42
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-23-5
22.99
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
sodium
Na
MFCD00085307
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
CHEBI:26708
[Na]

Specifications

White
99+% (Metals basis)
MFCD00085307
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
Air and moisture sensitive
Sodium
7440-23-5
Na
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
[Na]
22.99
CHEBI:26708
Dispersion
Videos
Safety and Handling

Safety and Handling

P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H260-H314-H335

missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: I

missing translation for 'einecsNumber' : 231-132-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only