missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dithionite, Extra Pure, SLR, Fisher Chemical™

Product Code. 10539580
Click to view available options
Quantity:
500 g
3 kg
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Sodium Dithionite, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.096
MDL Number 11640
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
PubChem CID 24489
ChEBI CHEBI:66870
SMILES [O-]S(=O)S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 300°C
Color White
pH 8 to 9.5
Physical Form Powder/Solid
Quantity 3 kg
Formula Weight 174.1g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Sodium hydrosulfite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Self-heating substance Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H251-Self-heating: may catch fire.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P264-Wash face, hands and anyA§ thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.