missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dodecyl sulfate, ≥99.5% alkyl sulfate, MP Biomedicals™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Assay Percent Range | >99.5% |
Quantity | 500 mL |
IUPAC Name | sodium dodecyl sulfate |
Formula Weight | 288.38 |
Percent Purity | ≥99.5% (alkyl sulfate) |
Chemical Name or Material | Sodium dodecyl sulfate, alkyl sulfate |
Safety and Handling
Product Identifier
- Sodium Dodecyl Sulfate, 20% Solution
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : sodium dodecyl sulphate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'