missing translation for 'onlineSavingsMsg'
Learn More
Sodium Dodecyl Sulfate (SDS), ≥95%, Thermo Scientific Chemicals Product Code.: 15865308

Sodium Dodecyl Sulfate (SDS), ≥95%, Thermo Scientific Chemicals

Product Code. 15865308 Shop All Thermo Scientific Products
1 kg, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Cardboard Carton
Plastic bottle
This item is not returnable. View return policy

Product Code. 15865308

Brand: Thermo Scientific J18220.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate,n to Dodecyl sulfate sodium salt
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Physical Form Crystalline Powder
Melting Point ∼206°C
Quantity 1 kg
Packaging Plastic bottle
Linear Formula CH3(CH2)11OSO3Na
UN Number UN2926
Beilstein 3599286
IUPAC Name sodium;dodecyl sulfate
Formula Weight 288.38
Percent Purity ≥95%
Chemical Name or Material Sodium Dodecyl Sulfate (SDS)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number232215 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog NumberJ18220.A1,J18220.36,J18220.A7
Lot Number242192 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ18220.A1,J18220.36,J18220.A7
Lot Number200082 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ18220.A1,J18220.36,J18220.A7
Lot Number231190 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberJ18220.A1,J18220.36,J18220.A7
Lot Number232201 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberJ18220.A1,J18220.36,J18220.A7
compliance-icons compliance-icons
Product Identifier
  • Sodium dodecyl sulfate, Thermo Scientific
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium lauryl sulphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Dodecyl Sulfate (SDS), ≥95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.