missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™

For protein extraction procedures and SDS polyacrylamide gels

£74.35 - £783.00

Chemical Identifiers

Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate, SDS
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10593335
View Documents
Fisher BioReagents
BP166-100
100 g Glass Bottle
£74.35
100g
Please to purchase this item. Need a web account? Register with us today!
10356463
View Documents
Fisher BioReagents
BP166-500
500 g Poly Bottle
£208.00
500g
Please to purchase this item. Need a web account? Register with us today!
10593355
View Documents
Fisher BioReagents
BP166-5
5 kg Poly Pail
£783.00
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For protein extraction procedures and SDS polyacrylamide gels

  • DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
  • SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
  • SDS denatures proteins by disrupting the non-covalent bonds of the protein
  • Functionally tested for use in electrophoresis
  • Intended for research use only; not for human or animal diagnostic purposes
Specifications

Chemical Identifiers

C12H25NaO4S
MFCD00036175
Sodium Lauryl Sulfate, SDS
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
sodium dodecyl sulfate

Specifications

0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm
151-21-3, 7647-14-5, 7757-82-6
206°C
Pass Test
C12H25NaO4S
MFCD00036175
Sodium Lauryl Sulfate, SDS
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
≥99% (≥98% C12-OH)
(as Fatty Alcohol Sulfate) >=99%
White
Solid
8.5 to 10
(as Fatty Alcohol Sulfate) >=99%
CH3(CH2)10CH2OSO3Na
15, 8768
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
DNase- and RNase-Free
Sodium Dodecyl Sulfate
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

Recommended Storage : RT

Product Identifier
  • Dodecyl sulfate sodium salt

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.