missing translation for 'onlineSavingsMsg'
Learn More

Sodium Edetate, Reagecon™
BIOPHARMA

Product Code. 11464809 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 11464809

Brand: Reagecon 3005900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 64-02-8
Molecular Formula C10H12N2Na4O8
Quantity 1 L
Synonym sodium edetate, edetate sodium, ethylenediaminetetraacetic acid tetrasodium salt, sequestrene, edta tetrasodium, sodium edta, tetrasodium edetate, aquamollin, complexone, irgalon
InChI Key UEUXEKPTXMALOB-UHFFFAOYSA-J
Molecular Weight (g/mol) 380.17
PubChem CID 6144
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.